missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(3-aminopropyl)-1,3-propanediamine-d24, CDN

Product Code. 30328368
Change view
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Product Code. Quantity unitSize
30328368 0.05 g 0.05g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30328368

Brand: CDN CDND61180.05G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Bis(3-aminopropyl)-1,3-propanediamine-D24
CAS 1219794-67-8
Molecular Formula C9 D24 N4
Quantity 0.05 g
Synonym N,N'-Bis(3-aminopropyl)-1,3-propanediamine-d24, N1,N3-Bis(3-aminopropyl-N3,N3,1,1,2,2,3,3-d8)-1,3-propanediamine-N1,N3,1,1,2,2,3,3-d8, Tripropylenetetramine-d24, 1,11-Diamino-4,8-diazaundecane-N1,N1,N11,N11,1,1,2,2,3,3,4,5,5,6,6,7,7,8,9,9,10,10,11,11-d24, Norspermine-d24
InChI Formula InChI=1 S/C9H24N4/c10-4-1-6-12-8-3-9-13-7-2-5-11/h12-13 H,1-11H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2/hD6
SMILES [2 H]N([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N([2 H])[2 H]
IUPAC Name N,N',N',1,1,2,2,3,3-nonadeuterio-N-[1,1,2,2,3,3-hexadeuterio-3-[deuterio-[1,1,2,2,3,3-hexadeuterio-3-(dideuterioamino)propyl]amino]propyl]propane-1,3-diamine
Molecular Weight (g/mol) 212.462
Formula Weight 212.351 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.