missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%

Product Code. 10494175 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10494175 50 g Glass bottle 50g
10001801 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10494175 Supplier Thermo Scientific Acros Supplier No. 345180500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4439-20-7 | C6H16N2O2 | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4439-20-7
Molecular Formula C6H16N2O2
Molecular Weight (g/mol) 148.2
MDL Number MFCD00002842
InChI Key GFIWSSUBVYLTRF-UHFFFAOYSA-N
Synonym n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol
PubChem CID 78179
IUPAC Name 2-[2-(2-hydroxyethylamino)ethylamino]ethanol
SMILES C(CNCCO)NCCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 92°C to 99°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (HOCH2CH2NHCH2-)2
Quantity 50 g
Formula Weight 148.2
Percent Purity 95%
Physical Form Chunks and/or Powder
Chemical Name or Material N, N'-Bis(2-hydroxyethyl)ethylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • N,N'-Bis(2-hydroxyethyl)ethylenediamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.