missing translation for 'onlineSavingsMsg'
Learn More

N,N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid, MP Biomedicals™

Product Code. 11394149 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394149 250 g 250g
11384149 100 g 100g
11304159 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394149 Supplier MP Biomedicals Supplier No. 0210092783
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10191-18-1 | C6H15NO5S | 213.25 g/mol

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N, N-Bis(2-hydroxyethyl)taurine, N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 0.9961
Formula Weight 239.23
Chemical Name or Material N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.