missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-chloroethyl)-p-toluenesulfonamide, Technical Grade 90%, TRC

Product Code. 30344151
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30344151 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30344151

Brand: Toronto Research Chemicals TRCB419010250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Bis(2-chloroethyl)-p-toluenesulfonamide
CAS 42137-88-2
Molecular Formula C11 H15 Cl2 N O2 S
Quantity 250 mg
Synonym Benzenesulfonamide, N,N-bis(2-chloroethyl)-4-methyl-, p-Toluenesulfonamide, N,N-bis(2-chloroethyl)- (6CI,7CI), N,N-Bis(2-chloroethyl)-4-methylbenzenesulfonamide, N,N-Bis(2-chloroethyl)-4-methylbenzensulfonamide, N,N-Bis(2-chloroethyl)-p-toluenesulfonamide, NSC 49364
InChI Formula InChI=1S/C11H15Cl2NO2S/c1-10-2-4-11(5-3-10)17(15,16)14(8-6-12)9-7-13/h2-5H,6-9H2,1H3
SMILES Cc1ccc(cc1)S(=O)(=O)N(CCCl)CCCl
IUPAC Name N,N-bis(2-chloroethyl)-4-methylbenzenesulfonamide
Molecular Weight (g/mol) 296.2133
Formula Weight 295.0201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.