missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(2-aminoethyl)-1,3-propanediamine, 96%

Product Code. 15461088
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461088 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15461088 Supplier Thermo Scientific Alfa Aesar Supplier No. H54950.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4741-99-5 | C7H20N4 | 160.265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4741-99-5
Density 0.96
Boiling Point 142°C to 145°C
Flash Point 100°C (212°F)
Molecular Formula C7H20N4
Linear Formula H2NCH2CH2NH(CH2)3NHCH2CH2NH2
Refractive Index 1.494
MDL Number MFCD00008174
Quantity 1 g
UN Number UN2735
Beilstein 1742732
Synonym n,n'-bis 2-aminoethyl-1,3-propanediamine, 1,4,8,11-tetraazaundecane, 2,3,2-tetramine, n,n'-bis 2-aminoethyl propane-1,3-diamine, 1,4,8,11-tetrazaundecane, ethylenetrimethyleneethylenetetramine, 1,3-propanediamine, n,n'-bis 2-aminoethyl, 1,9-diamino-3,7-diazanonane, n1,n1'-propane-1,3-diyl bis ethane-1,2-diamine, 3,7-diazanonane-1,9-diamine
InChI Key UWMHHZFHBCYGCV-UHFFFAOYSA-N
SMILES C(CNCCN)CNCCN
IUPAC Name N,N'-bis(2-aminoethyl)propane-1,3-diamine
Molecular Weight (g/mol) 160.265
PubChem CID 78479
ChEBI CHEBI:30348
Formula Weight 160.26
Percent Purity 97%
Chemical Name or Material N,N'-Bis(2-aminoethyl)-1,3-propanediamine
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.