missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride, Mikromol™
Description
N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride |
| CAS | 63434-11-7 |
| Analyte or Component Names | N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H38 N2 O4 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Verapamil Hydrochloride Imp. A (EP), Verapamil Imp. A (EP), N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride, Verapamil Hydrochloride Imp. A (EP) as Dihydrochloride, Verapamil Hydrochloride Impurity A as Dihydrochloride, Verapamil Impurity A as Dihydrochloride |
| SMILES | Cl.Cl.COc1ccc(CCN(C)CCCN(C)CCc2ccc(OC)c(OC)c2)cc1OC |
| Molecular Weight (g/mol) | 503.51 |
| Quantity | 25 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?