missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,beta-Trimethyl-10H-pyrido[3,2-b][1,4]benzothiazine-10-ethanamine Hydrochloride (beta-Isothipendyl Hydrochloride), Mikromol™
Description
N,N,beta-Trimethyl-10H-pyrido[3,2-b][1,4]benzothiazine-10-ethanamine Hydrochloride (beta-Isothipendyl Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | beta-Isothipendyl Hydrochloride |
| CAS | 132962-42-6 |
| Analyte or Component Names | beta-Isothipendyl Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 N3 S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 10H-Pyrido[3,2-b][1,4]benzothiazine-10-ethanamine, N,N,β-trimethyl-, hydrochloride (1:1), 10H-Pyrido[3,2-b][1,4]benzothiazine, 10-(2-dimethylamino-1-methylethyl)-, monohydrochloride (6CI), N,N,β-Trimethyl-10H-pyrido[3,2-b][1,4]benzothiazine-10-ethanamine Hydrochloride, β-Isothipendyl Hydrochloride |
| InChI Formula | InChI=1S/C16H19N3S.ClH/c1-12(11-18(2)3)19-13-7-4-5-8-14(13)20-15-9-6-10-17-16(15)19;/h4-10,12H,11H2,1-3H3;1H |
| SMILES | Cl.CC(CN(C)C)N1c2ccccc2Sc3cccnc13 |
| IUPAC Name | N,N-dimethyl-2-pyrido[3,2-b][1,4]benzothiazin-10-ylpropan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?