missing translation for 'onlineSavingsMsg'
Learn More

N-(N-Benzyloxycarbonyloxy)succinimide, TRC

Product Code. 30333932
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333932 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333932 Supplier Toronto Research Chemicals Supplier No. TRCB28627525G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(N-Benzyloxycarbonyloxy)succinimide
CAS 13139-17-8
Molecular Formula C12H11NO5
Quantity 25 g
Synonym 1-[[(Benzyloxy)carbonyl]oxy]-2,5-pyrrolidinedione, 1-[[(Phenylmethoxy)carbonyl]oxy]-2,5-pyrrolidinedione, 1-[[[(Phenylmethyl)oxy]carbonyl]oxy]-2,5-pyrrolidinedione, Benzyl 2,5-dioxopyrrolid-1-yl Carbonate, Benzyl 2,5-dioxopyrrolidin-1-yl Carbonate, Benzyl N-succinimidyl Carbonate, Benzyl Succinimidyl Carbonate, Benzyloxycarbonyloxysuccinimide, Cbz-OSu, N-(Benzyloxycarbonyloxy)succinimide
InChI Formula InChI=1S/C12H11NO5/c14-10-6-7-11(15)13(10)18-12(16)17-8-9-4-2-1-3-5-9/h1-5H,6-8H2
SMILES O=C(OCc1ccccc1)ON2C(=O)CCC2=O
IUPAC Name benzyl (2,5-dioxopyrrolidin-1-yl) carbonate
Molecular Weight (g/mol) 249.22
Formula Weight 249.0637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.