missing translation for 'onlineSavingsMsg'
Learn More

N,N,3,5-Tetramethylaniline, 98%

Product Code. 11357316
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357316 5 g 5g
11367316 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357316 Supplier Thermo Scientific Alfa Aesar Supplier No. L09266.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4913-13-7 | C10H15N | 149.237 g/mol

N,N,3,5-Tetramethylaniline was used in rapid quenching of 1,8-dihydroxyanthraquinone (DHAQ). It was also used as a catalyst in the polymeric synthesis of glycol methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,3,5-Tetramethylaniline was used in rapid quenching of 1,8-dihydroxyanthraquinone (DHAQ). It was also used as a catalyst in the polymeric synthesis of glycol methacrylate.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4913-13-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008307
InChI Key NBFRQCOZERNGEX-UHFFFAOYSA-N
Synonym benzenamine, n,n,3,5-tetramethyl, n,n-dimethyl-3,5-xylidine, 3,5-xylidine, n,n-dimethyl, 3,5-dimethylphenyl dimethylamine, acmc-209kew, 5-dimethylamino-m-xylene, 3,5,n,n-tetramethylaniline, n,n-dimethyl-sym-m-xylidine, n,n,3,5-tetramethyl aniline, n,n-3,5-tetramethylbenzenamine
PubChem CID 78625
IUPAC Name N,N,3,5-tetramethylaniline
SMILES CC1=CC(=CC(=C1)N(C)C)C

Specifications

Density 0.913
Quantity 5 g
Odor Recognizable
Refractive Index 1.544
UN Number UN2810
Beilstein 1933736
Boiling Point 226°C to 228°C
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 149.24
Percent Purity 98%
Chemical Name or Material N,N,3,5-Tetramethylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.