missing translation for 'onlineSavingsMsg'
Learn More

N,N,2,4,6-Pentamethylaniline, 98%, Thermo Scientific Chemicals

Product Code. 11453204
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453204 100 g 100g
11443204 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453204 Supplier Thermo Scientific Alfa Aesar Supplier No. A10561.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13021-15-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00010596
InChI Key JZBZLRKFJWQZHU-UHFFFAOYSA-N
Synonym n,n,2,4,6-pentamethylbenzenamine, 2-dimethylamino mesitylene, n,n-dimethylmesidine, benzenamine,n,n,2,4,6-pentamethyl, 2-dimethylaminomesitylene, dimethylaminomesitylen, pubchem3219, acmc-209bhq, n,n-2,4,6-pentamethylaniline, 2,4,6,n,n-pentamethylbenzenamine
PubChem CID 139365
IUPAC Name N,N,2,4,6-pentamethylaniline
SMILES CC1=CC(=C(C(=C1)C)N(C)C)C

Specifications

Density 0.907
Boiling Point 213°C to 215°C
Flash Point 78°C (174°F)
Refractive Index 1.521
Quantity 100 g
Formula Weight 163.27
Percent Purity 98%
Chemical Name or Material N,N,2,4,6-Pentamethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.