missing translation for 'onlineSavingsMsg'
Learn More

N,N,2,2-Tetramethyl-1,3-propanediamine, 97%, Thermo Scientific™

Product Code. 10351180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351180 25 g Glass bottle 25g
10863381 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351180 Supplier Thermo Scientific Acros Supplier No. 210870250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 53369-71-4
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.23
InChI Key ULDIVZQLPBUHAG-UHFFFAOYSA-N
Synonym n,n,2,2-tetramethyl-1,3-propanediamine, 1,3-propanediamine, n,n,2,2-tetramethyl, n,n,2,2-tetramethylpropane-1,3-diamine, n,n-dimethylneopentanediamine, 3-amino-2,2-dimethylpropyl dimethylamine, n1,n1,2,2-tetramethylpropane-1,3-diamine, 1,3-propanediamine, n1,n1,2,2-tetramethyl, acmc-209l6k, ksc597o9f, uldivzqlpbuhag-uhfffaoysa
PubChem CID 104477
IUPAC Name N',N',2,2-tetramethylpropane-1,3-diamine
SMILES CC(C)(CN)CN(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 155.0°C to 158.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH2C(CH3)2CH2NH2
Refractive Index 1.4355
Quantity 25 g
Beilstein 04,III,596
Specific Gravity 0.82
Formula Weight 130.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N,N,2,2-Tetramethyl-1,3-propanediamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N,2,2-Tetramethyl-1,3-propanediamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.