missing translation for 'onlineSavingsMsg'
Learn More

N,N'-1,2-Ethanediylbis[N-methy-b-alanine Diethyl Ester], TRC

Product Code. 30380403
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380403 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380403 Supplier Toronto Research Chemicals Supplier No. TRCE9176705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-1,2-Ethanediylbis[N-methy-beta-alanine Diethyl Ester]
CAS 42434-14-0
Molecular Formula C14H28N2O4
Quantity 5 g
InChI Formula InChI=1S/C14H28N2O4/c1-5-19-13(17)7-9-15(3)11-12-16(4)10-8-14(18)20-6-2/h5-12H2,1-4H3
SMILES O=C(OCC)CCN(C)CCN(C)CCC(=O)OCC
IUPAC Name diethyl 3,3'-(ethane-1,2-diylbis(methylazanediyl))dipropionate
Molecular Weight (g/mol) 288.38
Formula Weight 288.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.