missing translation for 'onlineSavingsMsg'
Learn More

N,N'-1,2-ethanediylbis-Carbamodithioic acid C,C'-dimethyl ester, TRC

Product Code. 30134512
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134512 5 mg 5mg
30132411 10 mg 10mg
17982095 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134512 Supplier Toronto Research Chemicals Supplier No. TRCE5013205MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20721-48-6
Molecular Formula C6 H12 N2 S4
Molecular Weight (g/mol) 240.4329
Synonym Carbamic acid, ethylenebis[dithio-, dimethyl ester (6CI,7CI,8CI), Carbamodithioic acid, 1,2-ethanediylbis-, dimethyl ester (9CI), C,C'-Dimethyl N,N'-1,2-ethanediylbis[carbamodithioate], S,S'-Dimethylxanthogenethylenebisdithiocarbamate
IUPAC Name methyl N-[2-(methylsulfanylcarbothioylamino)ethyl]carbamodithioate
SMILES CSC(=S)NCCNC(=S)SC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H12N2S4/c1-11-5(9)7-3-4-8-6(10)12-2/h3-4H2,1-2H3,(H,7,9)(H,8,10)
Formula Weight 239.9883
Percent Purity >95%
Chemical Name or Material N,N'-1,2-ethanediylbis-Carbamodithioic acid C,C'-dimethyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.