missing translation for 'onlineSavingsMsg'
Learn More

n-Myc Antibody, Novus Biologicals™

Product Code. 18358844 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358844 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358844 Supplier Novus Biologicals Supplier No. NBP313281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

n-Myc Polyclonal antibody specifically detects n-Myc in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen n-Myc
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS, 20% Glycerol (pH7)
Gene Alias BHLHE37, bHLHe37N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, MODED, neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene, N-myc, NMYCneuroblastoma MYC oncogene, ODED, oncogene NMYC, pp65/67, v-myc avian myelocytomatosis viral related oncogene, neuroblastoma derived, v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian)
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human n-Myc. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4613
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.