missing translation for 'onlineSavingsMsg'
Learn More

N-Methylsulfanilic acid, 98+%, Thermo Scientific™

Product Code. 11394414
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11394414

Brand: Thermo Scientific Alfa Aesar L01016.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24447-99-2
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00014732
InChI Key QRAXZXPSAGQUNP-UHFFFAOYSA-N
Synonym 4-methylamino benzenesulfonic acid, 4-methylamino benzenesulphonic acid, n-methylsulphanilic acid, n-methylsulfanilic acid, benzenesulfonic acid,4-methylamino, n-methylaniline-p-sulfonic acid, maybridge1_004198, n-methylsulfanilic acid, 98+%
PubChem CID 90505
IUPAC Name 4-(methylamino)benzenesulfonic acid
SMILES CNC1=CC=C(C=C1)S(=O)(=O)O

Specifications

Melting Point ∽260°C (decomposition)
Quantity 25 g
Beilstein 2094129
Formula Weight 187.21
Percent Purity ≥98%
Chemical Name or Material N-Methylsulfanilic acid
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado