missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrolidone, Mikromol™

Product Code. 18001360
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 18001360

Brand: Mikromol MM3004.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Methylpyrrolidone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methyl-2-pyrrolidon
CAS 872-50-4
Analyte or Component Names 1-Methyl-2-pyrrolidon
Shipping Condition Room Temperature
Molecular Formula C5 H9 N O
Certifications/Compliance ISO 17025
Synonym N-Methylpyrrolidone, 1-Methylpyrrolidin-2-one
InChI Formula InChI=1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3
SMILES CN1CCCC1=O
IUPAC Name 1-methylpyrrolidin-2-one
Molecular Weight (g/mol) 99.13
Quantity 250 mg
Formula Weight 99.0684 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.