missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrole, TRC

Product Code. 30483048
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483048 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483048 Supplier Toronto Research Chemicals Supplier No. TRCM327015250ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methylpyrrole
CAS 96-54-8
Molecular Formula C5H7N
Quantity 250 mL
Synonym 1-Methyl-1H-pyrrole, 1-Methylpyrrole, NSC 65440
InChI Formula InChI=1S/C5H7N/c1-6-4-2-3-5-6/h2-5H,1H3
SMILES Cn1cccc1
Molecular Weight (g/mol) 81.12
Formula Weight 81.0578
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.