missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrole, 99+%

Product Code. 10457900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10457900 100 g Glass bottle 100g
10234890 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10457900 Supplier Thermo Scientific Acros Supplier No. 127601000
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 96-54-8 | C5H7N | 81.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-54-8
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.12
MDL Number MFCD00005345
InChI Key OXHNLMTVIGZXSG-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole, n-methylpyrrole, n-methyl pyrrole, 1h-pyrrole, 1-methyl, pyrrole, 1-methyl, methylpyrrole, n-methylpyrrol, 1-methylpyrrol, unii-ng5gpn98zg, ccris 2934
PubChem CID 7304
IUPAC Name 1-methylpyrrole
SMILES CN1C=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -57°C
Color Yellow to Brown
Density 0.9100g/mL
Boiling Point 111°C to 114°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4875
Quantity 100 g
Beilstein 20, 163
Specific Gravity 0.91
Solubility Information Solubility in water: 13g/L water (20°C). Other solubilities: soluble in alcohol,miscible with most usual organic solents
Viscosity 0.7 mPa.s (20°C)
Formula Weight 81.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N-Methylpyrrole, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methylpyrrole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.