missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpropargylamine, 98+%

Product Code. 10248364 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10248364 5 g Glass bottle 5g
10594578 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10248364 Supplier Thermo Scientific Acros Supplier No. 165540050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35161-71-8 | C4H7N | 69.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35161-71-8
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.11
MDL Number MFCD00008573
InChI Key HQFYIDOMCULPIW-UHFFFAOYSA-N
Synonym n-methylpropargylamine, 2-propyn-1-amine, n-methyl, methyl-prop-2-ynyl-amine, 3-methylamino-1-propyne, n-methylpropyn-2-ylamine, n-methyl-n-prop-2-ynylamine, n-methyl-2-propyn-1-amine, n-methyl-n-propargylamine, methyl prop-2-yn-1-yl amine, 2-propyn-1-amine,n-methyl-9ci
PubChem CID 96160
IUPAC Name N-methylprop-2-yn-1-amine
SMILES CNCC#C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Density 0.8300g/mL
Boiling Point 82°C to 84°C
Flash Point 1°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HC∽CCH2NHCH3
Refractive Index 1.4332
Quantity 5 g
Beilstein 04,228
Specific Gravity 0.83
Formula Weight 69.11
Percent Purity 98+%
Chemical Name or Material N-Methylpropargylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methylpropargylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N-Methylpropargylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.