missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmorpholine un2535-ii, 500 mL, MP Biomedicals™

Product Code. 11412402 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412402 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412402 Supplier MP Biomedicals Supplier No. 0215166490
  • ONLINE EXCLUSIVE PRICE
    £318.00
    £298.20 / 500mL
    Save £19.80
    6% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

A solvent for peptide and protein synthesis and sequencing and starting material for preparing N-methylmorpholine N-oxide

Specifications

CAS 109-02-4
Melting Point -66°C
Density 0.92 g/mL at 25 °C
Boiling Point 115°C to 116°C (at 750 mmHg)
Flash Point 14 °C
Molecular Formula C5H11NO
Quantity 500 mL
Synonym Methylmorpholine, 4-methylmorpholine, NMM
InChI Key SJRJJKPEHAURKC-UHFFFAOYSA-N
SMILES CN1CCOCC1
IUPAC Name 4-methylmorpholine
Molecular Weight (g/mol) 101.15
Vapor Density >1
Vapor Pressure 18 mmHg at 20 °C
Physical Form Liquid
Show More Show Less

For manufacturing use only, not for direct human use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.