missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmorpholine (Peptide Synthesis), Fisher BioReagents

Product Code. 10688393
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10688393 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10688393 Supplier Fisher BioReagents Supplier No. BP610500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
N-Methylmorpholine is used in peptide synthesis protocols.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Methylmorpholine
CAS 109-02-4
Boiling Range 2 max.
Melting Point -65°C
Color Colorless
Density 0.920g/cm³
pH 10.6
Boiling Point 115°C
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Molecular Formula C5H11NO
MDL Number MFCD00006175
Quantity 500 mL
Merck Index 15, 6362
Synonym n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin
InChI Key SJRJJKPEHAURKC-UHFFFAOYSA-N
SMILES CN1CCOCC1
IUPAC Name 4-methylmorpholine
Molecular Weight (g/mol) 101.15
PubChem CID 7972
Formula Weight 101.15
Vapor Pressure 22.5mmHg at 20°C
Percent Purity ≥99.0%
Grade Peptide Synthesis
Physical Form Liquid
Chemical Name or Material N-Methylmorpholine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Methylmorpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.