missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmaleimide, 98+%

Product Code. 11310044
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310044 25 g 25g
11300044 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310044 Supplier Thermo Scientific Alfa Aesar Supplier No. L00437.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 930-88-1 | C5H5NO2 | 111.1 g/mol

N-Methylmaleimide is an electron deficient olefin that acts as a thiol-blocking reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylmaleimide is an electron deficient olefin that acts as a thiol-blocking reagent.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-88-1
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00005508
InChI Key SEEYREPSKCQBBF-UHFFFAOYSA-N
Synonym n-methylmaleimide, 1-methyl-1h-pyrrole-2,5-dione, maleimide, n-methyl, n-methylmaleinimide, 1h-pyrrole-2,5-dione, 1-methyl, n-methyl maleimide, unii-p0tfz8r21y, p0tfz8r21y, 1-methyl-pyrrole-2,5-dione, 1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione
PubChem CID 70261
IUPAC Name 1-methylpyrrole-2,5-dione
SMILES CN1C(=O)C=CC1=O

Specifications

Melting Point 94°C to 96°C
Odor Odorless
Quantity 25 g
UN Number UN2928
Beilstein 108550
Solubility Information Slightly soluble in water.
Formula Weight 111.1
Percent Purity ≥98%
Chemical Name or Material N-Methylmaleimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.