Learn More
N-Methylisobutylamine, 98+%, Thermo Scientific Chemicals
£23.40 - £69.30
Chemical Identifiers
CAS | 625-43-4 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.166 |
MDL Number | MFCD00015043 |
InChI Key | QKYWADPCTHTJHQ-UHFFFAOYSA-N |
Synonym | n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin |
PubChem CID | 12249 |
IUPAC Name | N,2-dimethylpropan-1-amine |
SMILES | CC(C)CNC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11370218
|
Thermo Scientific Alfa Aesar
042947.03 |
1 g |
£23.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11360218
|
Thermo Scientific Alfa Aesar
042947.06 |
5 g |
£69.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Methylisobutylamine is used as an pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
625-43-4 | |
87.166 | |
QKYWADPCTHTJHQ-UHFFFAOYSA-N | |
12249 | |
CC(C)CNC |
C5H13N | |
MFCD00015043 | |
n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin | |
N,2-dimethylpropan-1-amine |
Specifications
625-43-4 | |
78°C | |
C5H13N | |
UN2733 | |
Miscible with water. | |
CC(C)CNC | |
87.166 | |
87.16 | |
Liquid |
0.73 g/mL | |
Amine-like | |
MFCD00015043 | |
n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin | |
QKYWADPCTHTJHQ-UHFFFAOYSA-N | |
N,2-dimethylpropan-1-amine | |
12249 | |
≥98% | |
N-Methylisobutylamine |
Safety and Handling
GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 210-893-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only