missing translation for 'onlineSavingsMsg'
Learn More

N-Methyliminodiacetic acid, 99%

Product Code. 11337755
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11337755

Brand: Thermo Scientific Alfa Aesar L02589.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4408-64-4 | C5H9NO4 | 147.13 g/mol

N-Methyliminodiacetic acid is used to prepare bis-(1H-benzoimidazol-2-ylmethyl)-methyl-amine by condensation with benzene-1,2-diamine. It is involved in Suzuki cross-coupling with N-methyliminodiacetic acid (MIDA) boronates. It acts as a reagent utilized for protecting boronic acids and as a chelating agent for various metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyliminodiacetic acid is used to prepare bis-(1H-benzoimidazol-2-ylmethyl)-methyl-amine by condensation with benzene-1,2-diamine. It is involved in Suzuki cross-coupling with N-methyliminodiacetic acid (MIDA) boronates. It acts as a reagent utilized for protecting boronic acids and as a chelating agent for various metals.

Solubility
Soluble in water. Slightly soluble in dimethyl sulfoxide.

Notes
Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4408-64-4
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00004284
InChI Key XWSGEVNYFYKXCP-UHFFFAOYSA-N
Synonym n-methyliminodiacetic acid, 2,2'-methylazanediyl diacetic acid, methyliminodiacetic acid, mida, methylimidodiacetic acid, n-methyliminodiaceticacid, glycine, n-carboxymethyl-n-methyl, methylimino diacetic acid, acetic acid, methylimino di, carboxymethyl methyl amino acetic acid
PubChem CID 20441
IUPAC Name 2-[carboxymethyl(methyl)amino]acetic acid
SMILES CN(CC(=O)O)CC(=O)O

Specifications

Melting Point ∽220°C (decomposition)
Quantity 5 g
Beilstein 1765449
Solubility Information Soluble in water. Slightly soluble in dimethyl sulfoxide.
Formula Weight 147.13
Percent Purity 99%
Chemical Name or Material N-Methyliminodiacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.