Learn More
N-Methylformanilide, 99%
Description
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.
Solubility
Immisible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 9°C to 13°C |
| Density | 1.095 |
| Boiling Point | 242°C to 244°C |
| Flash Point | 126°C (258°F) |
| Refractive Index | 1.561 |
| Quantity | 2500 g |
| Beilstein | 636496 |
| Solubility Information | Immisible with water. |
| Formula Weight | 135.17 |
| Percent Purity | 99% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.