missing translation for 'onlineSavingsMsg'
Learn More

N-Methylethylenediamine, 95%

Product Code. 11484793
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484793 25 g 25g
11474793 5 g 5g
11494793 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484793 Supplier Thermo Scientific Alfa Aesar Supplier No. A12165.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-81-9 | C3H12N2 | 76.14 g/mol

N-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-81-9
Molecular Formula C3H12N2
Molecular Weight (g/mol) 76.14
MDL Number MFCD00008165
InChI Key KFIGICHILYTCJF-UHFFFAOYSA-P
Synonym n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
PubChem CID 8014
IUPAC Name N'-methylethane-1,2-diamine
SMILES C[NH2+]CC[NH3+]

Specifications

Density 0.851
Boiling Point 114°C to 116°C
Flash Point 41°C (105°F)
Odor Amine-like
Linear Formula H2NCH2CH2NHCH3
Refractive Index 1.4395
Quantity 25 g
UN Number UN2734
Beilstein 878141
Sensitivity Air sensitive; Hygroscopic
Formula Weight 74.13
Percent Purity 95%
Chemical Name or Material N-Methylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.