missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldopamine hydrochloride, 95%

Product Code. 15473099
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473099 5 g 5g
15463099 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473099 Supplier Thermo Scientific Alfa Aesar Supplier No. J60306.06
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dopamine agonist | CAS: 62-32-8 | C9H14ClNO2 | 203.67 g/mol

Andregenic and dopamine receptor agonist; active metabolite of Ibopramine. Also, it is used as internal standard in chromatographic analysis of catecholamine's.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Andregenic and dopamine receptor agonist; active metabolite of Ibopramine. Also, it is used as internal standard in chromatographic analysis of catecholamine′s.

Solubility
Soluble in methanol, water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-32-8
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.67
MDL Number MFCD00035074
InChI Key JCDRZCWRRLKLTB-UHFFFAOYSA-N
Synonym n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride
PubChem CID 86470
IUPAC Name hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride
SMILES [H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in methanol,water.
Formula Weight 203.67
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Methyldopamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.