missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 105-59-9
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00002848
InChI Key CRVGTESFCCXCTH-UHFFFAOYSA-N
Synonym methyldiethanolamine, n-methyliminodiethanol, n-methyldiethanolamine, 2,2'-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, methyl diethanolamine, ethanol, 2,2'-methylimino bis, n-methyldiethanolimine, n-methylaminodiglycol, 2,2'-methylimino diethanol
PubChem CID 7767
SMILES CN(CCO)CCO
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

105-59-9
119.16
CRVGTESFCCXCTH-UHFFFAOYSA-N
7767
C5H13NO2
MFCD00002848
methyldiethanolamine, n-methyliminodiethanol, n-methyldiethanolamine, 2,2'-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, methyl diethanolamine, ethanol, 2,2'-methylimino bis, n-methyldiethanolimine, n-methylaminodiglycol, 2,2'-methylimino diethanol
CN(CCO)CCO

Specifications

105-59-9
100.0
Colorless to Yellow
243°C
Authentic
C5H13NO2
1.4675 to 1.4695
04, 284
methyldiethanolamine, n-methyliminodiethanol, n-methyldiethanolamine, 2,2'-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, methyl diethanolamine, ethanol, 2,2'-methylimino bis, n-methyldiethanolimine, n-methylaminodiglycol, 2,2'-methylimino diethanol
CRVGTESFCCXCTH-UHFFFAOYSA-N
2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
7767
119.16
Liquid
99.0
-21°C
1.0380 g/mL
138°C
99% min. (GC)
(HOCH2CH2)2NCH3
MFCD00002848
1.038
Solubility aq. soln.: miscible. Other solubilities: miscible with benzene,soluble in most organic solvents (alcohol,esters,aceton and chlorinated hydrocarbons),soluble in diethylether and aliphatic hydrocarbons
CN(CCO)CCO
119.16
101 mPa.s (20°C)
99+%
N-Methyldiethanolamine, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:

Causes serious eye irritation.

GHS P Statement:

Wash face, hands and any exposed skin thoroughly after handling.

Wear protective gloves/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

If eye irritation persists: Get medical advice/attention.



WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 203-312-7

missing translation for 'rtecsNumber' : KL7525000

missing translation for 'tsca' : TSCA

Product Identifier
  • N-Methyldiethanolamine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only