missing translation for 'onlineSavingsMsg'
Learn More

N-Methylcyclohexylamine, 98%

Product Code. 11303297
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11303297 100 mL 100mL
11393287 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303297 Supplier Thermo Scientific Alfa Aesar Supplier No. L13872.AE
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-60-7 | C7H15N | 113.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 100-60-7
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00003832
InChI Key XTUVJUMINZSXGF-UHFFFAOYSA-N
Synonym n-methylcyclohexylamine, methylcyclohexylamine, n-cyclohexylmethylamine, cyclohexanamine, n-methyl, n-methyl-n-cyclohexylamine, methylamino cyclohexane, cyclohexylamine, n-methyl, cyclohexyl methyl amine, n-cyclohexyl-n-methylamine, unii-yh8pf4c1ow
PubChem CID 7514
ChEBI CHEBI:59018
IUPAC Name N-methylcyclohexanamine
SMILES CNC1CCCCC1

Tekniske data

Melting Point -8°C
Density 0.857
Boiling Point 149°C
Flash Point 35°C (95°F)
Refractive Index 1.456
Quantity 100 mL
UN Number UN2734
Beilstein 1523664
Sensitivity Air Sensitive
Formula Weight 113.2
Percent Purity 98%
Chemical Name or Material N-Methylcyclohexylamine
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.