missing translation for 'onlineSavingsMsg'
Learn More

N-(Methylcarbamoyl) Sulfanilamide-N4-acetate, TRC

Product Code. 30479775
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479775 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479775 Supplier Toronto Research Chemicals Supplier No. TRCM329335100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 18431-25-9
Molecular Formula C10H12N2O5S
Quantity 100 mg
Synonym Methyl (4-acetamidophenyl)sulfonylcarbamate
InChI Formula InChI=1S/C10H12N2O5S/c1-7(13)11-8-3-5-9(6-4-8)18(15,16)12-10(14)17-2/h3-6H,1-2H3,(H,11,13)(H,12,14)
SMILES COC(=O)NS(=O)(=O)c1ccc(NC(=O)C)cc1
IUPAC Name methyl N-(4-acetamidophenyl)sulfonylcarbamate
Molecular Weight (g/mol) 272.28
Formula Weight 272.0467
Chemical Name or Material N-(Methylcarbamoyl) Sulfanilamide-N4-acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.