missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzylamine, TRC

Product Code. 30134743
Change view
Click to view available options
Quantity:
5 mL
100 mL
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134743 100 mL 100mL
30007748 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134743 Supplier Toronto Research Chemicals Supplier No. TRCM288990100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-67-3
InChI Formula InChI=1S/C8H11N/c1-9-7-8-5-3-2-4-6-8/h2-6,9H,7H2,1H3
IUPAC Name N-methyl-1-phenylmethanamine
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
Chemical Name or Material N-Methylbenzylamine

Specifications

Quantity 100 mL
Synonym N-Methylbenzenemethanamine, N-Methyl-N-(phenylmethyl)amine, N-Methyl-N-benzylamine, N-Benzylmethylamine, N-Methyl-1-phenylmethanamine, N-(Phenylmethyl)methylamine, N-Benzyl-N-methylamine, NSC 8059;
SMILES CNCc1ccccc1
Formula Weight 121.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.