missing translation for 'onlineSavingsMsg'
Learn More

N-Methylazetidin-3-amine Dihydrochloride, TRC

Product Code. 30468494
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468494 500 mg 500mg
30480275 100 mg 100mg
30483624 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468494 Supplier Toronto Research Chemicals Supplier No. TRCM288718500MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136293-86-2
Molecular Formula C4H10N2 . 2HCl
Synonym 3-Azetidinamine, N-methyl-, hydrochloride (1:2) (ACI), 3-Azetidinamine, N-methyl-, dihydrochloride (9CI), 3-(Methylamino)azetidine dihydrochloride, N-Methyl-3-azetidinamine dihydrochloride, N-Methylazetidin-3-amine dihydrochloride
IUPAC Name N-methylazetidin-3-amine;dihydrochloride
SMILES Cl.Cl.CNC1CNC1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H10N2.2ClH/c1-5-4-2-6-3-4;;/h4-6H,2-3H2,1H3;2*1H
Formula Weight 158.0378
Chemical Name or Material N-Methylazetidin-3-amine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.