missing translation for 'onlineSavingsMsg'
Learn More

N-Methylaniline, 99%

Product Code. 10023223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10023223 2.5 kg Glass bottle 2.5kg
10408670 500 g Glass bottle 500g
10703891 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10023223 Supplier Thermo Scientific Acros Supplier No. 126270025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-61-8 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-61-8
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00008283
InChI Key AFBPFSWMIHJQDM-UHFFFAOYSA-N
Synonym methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
PubChem CID 7515
ChEBI CHEBI:15733
IUPAC Name N-methylaniline
SMILES CNC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -57.0°C
Color Brown to Yellow
Density 0.9870g/mL
Boiling Point 195.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NHCH3
Refractive Index 1.5694 to 1.5714
Quantity 2.5 kg
Beilstein 12, 135
Fieser 14,207
Merck Index 15, 6093
Specific Gravity 0.987
Solubility Information Solubility in water: 30g/l. Other solubilities: soluble in alcohol and ether,soluble in acetone,diethyl ether,ethanol and,chloroform
Formula Weight 107.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.