missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Zonisamide, TRC

Product Code. 30469745
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
Product Code. Quantity unitSize
30469745 250 mg 250mg
30464686 25 mg 25mg
2 options
This item is not returnable. View return policy

Product Code. 30469745

Brand: Toronto Research Chemicals TRCM338605250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68292-02-4
Formula Weight 226.0412
InChI Formula InChI=1S/C9H10N2O3S/c1-10-15(12,13)6-8-7-4-2-3-5-9(7)14-11-8/h2-5,10H,6H2,1H3
IUPAC Name 1-(1,2-benzoxazol-3-yl)-N-methylmethanesulfonamide
Molecular Formula C9H10N2O3S
Molecular Weight (g/mol) 226.25
SMILES CNS(=O)(=O)Cc1noc2ccccc12
Synonym N-Methyl-1,2-benzisoxazole-3-methanesulfonamide
Chemical Name or Material N-Methyl Zonisamide

Specifications

Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.