missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Omeprazole (Mixture of isomers with the methylated nitrogens of imidazole), TRC

Product Code. 17971805
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971805 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971805 Supplier Toronto Research Chemicals Supplier No. TRCM324500100MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 89352-76-1+784143-42-6
Molecular Formula C18H21N3O3S
Quantity 100 mg
Synonym 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1-methyl-1H-benzimidazole;
InChI Formula InChI=1S/2C18H21N3O3S/c1-11-9-19-15(12(2)17(11)24-5)10-25(22)18-20-14-8-13(23-4)6-7-16(14)21(18)3;1-11-9-19-15(12(2)17(11)24-5)10-25(22)18-20-14-7-6-13(23-4)8-16(14)21(18)3/h2*6-9H,10H2,1-5H3
SMILES O=S(CC1=C(C)C(OC)=C(C)C=N1)C(N2C)=NC3=C2C=CC(OC)=C3.O=S(CC4=C(C)C(OC)=C(C)C=N4)C5=NC6=C(N5C)C=C(OC)C=C6
Molecular Weight (g/mol) 359.44
Formula Weight 259.13
Percent Purity >95%
Chemical Name or Material N-Methyl Omeprazole (Mixture of isomers with the methylated nitrogens of imidazole)
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.