missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-trimethylsilylacetamide, 98%

Product Code. 11376786
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376786 10 g 10g
11366786 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376786 Supplier Thermo Scientific Alfa Aesar Supplier No. L04101.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7449-74-3 | C6H15NOSi | 145.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7449-74-3
Molecular Formula C6H15NOSi
Molecular Weight (g/mol) 145.277
MDL Number MFCD00040758
InChI Key QHUOBLDKFGCVCG-UHFFFAOYSA-N
Synonym n-methyl-n-trimethylsilyl acetamide, acetamide, n-methyl-n-trimethylsilyl, n-trimethylsilyl-n-methylacetamide, n-methyl-n-tms-acetamide, acmc-1bei6, qhuobldkfgcvcg-uhfffaoysa, n-methylacetamide, tms derivative, n-methyl-n-trimethylsilylacetamide, for gc derivatization gc
PubChem CID 81953
IUPAC Name N-methyl-N-trimethylsilylacetamide
SMILES CC(=O)N(C)[Si](C)(C)C

Specifications

Boiling Point 48°C to 49°C (11mmHg)
Quantity 10 g
UN Number UN1993
Beilstein 506975
Flash Point 48°C (118°F)
Formula Weight 145.28
Percent Purity 95%
Refractive Index 1.439
Sensitivity Moisture sensitive
Density 0.906
Chemical Name or Material N-Methyl-N-trimethylsilylacetamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.