missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-propargylbenzylamine, 97%

Product Code. 10128083 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10128083 25 g Glass Bottle 25g
10183220 1 g Glass Bottle 1g
10193220 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10128083 Supplier Thermo Scientific Acros Supplier No. 127530250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 555-57-7 | C11H13N | 159.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-57-7
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
MDL Number MFCD00008576
InChI Key DPWPWRLQFGFJFI-UHFFFAOYSA-N
Synonym pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
PubChem CID 4688
ChEBI CHEBI:7930
IUPAC Name N-benzyl-N-methylprop-2-yn-1-amine
SMILES CN(CC#C)CC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9400g/mL
Boiling Point 86°C to 88°C (4.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula C6H5CH2N(CH3)CH2C∽CH
Refractive Index 1.5213
Quantity 25 g
Beilstein 12, II, 548
Merck Index 15, 7142
Specific Gravity 0.94
Solubility Information Solubility in water: soluble. Other solubilities: miscibel in ethanol,acetone
Formula Weight 159.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methyl-N-propargylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methyl-N-propargylbenzylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.