missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-n-octylamine, 97+%

Product Code. 11339435
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339435 25 g 25g
11329435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339435 Supplier Thermo Scientific Alfa Aesar Supplier No. L18639.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2439-54-5 | C9H21N | 143.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2439-54-5
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.274
MDL Number MFCD00048927
InChI Key SEGJNMCIMOLEDM-UHFFFAOYSA-N
Synonym n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
PubChem CID 75538
IUPAC Name N-methyloctan-1-amine
SMILES CCCCCCCCNC

Specifications

Density 0.79
Boiling Point 68°C (8 mmHg)
Flash Point 63°C (145°F)
Linear Formula CH3(CH2)7NHCH3
Refractive Index 1.427
Quantity 25 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 143.27
Percent Purity 98%
Chemical Name or Material N-Methyl-n-octylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.