missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-nitroso-2-propanamine, TRC

Product Code. 30011625
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011625 250 mg 250mg
30005325 2.5 g 2.5g
30003450 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011625 Supplier Toronto Research Chemicals Supplier No. TRCM350800250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30533-08-5
Molecular Formula C4 H10 N2 O
Molecular Weight (g/mol) 102.14
Synonym 2-Propanamine, N-methyl-N-nitroso-, Ethylamine, N,1-dimethyl-N-nitroso- (6CI,7CI,8CI), N-Methyl-N-nitroso-2-propanamine, Isopropylmethylnitrosamine, Methylisopropylnitrosamine, N,1-Dimethyl-N-nitrosoethanamine, N-Methyl-N-nitrosoisopropylamine, NMiPA
IUPAC Name N-methyl-N-propan-2-ylnitrous amide
SMILES CC(C)N(C)N=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H10N2O/c1-4(2)6(3)5-7/h4H,1-3H3
Formula Weight 102.0793
Percent Purity >95%
Chemical Name or Material N-Nitroso-N-methyl-N-isopropylamine
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.