missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(6-morpholin-4-ylpyridin-2-yl)methyl]amine 90+%, Thermo Scientific™

Product Code. 10281893
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10281893 1 g Amber Glass Bottle 1g
10630404 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10281893 Supplier Thermo Scientific Maybridge Supplier No. CC38346DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-30-5
Molecular Formula C11H17N3O
Molecular Weight (g/mol) 207.28
MDL Number MFCD08435863
InChI Key CULRQHZBTPUFEB-UHFFFAOYSA-N
Synonym n-methyl-n-6-morpholin-4-ylpyridin-2-yl methyl amine, methyl 6-morpholin-4-yl pyridin-2-yl methyl amine, methyl 6-morpholin-4-yl 2-pyridyl methyl amine, 2-pyridinemethanamine,n-methyl-6-4-morpholinyl, n-methyl-1-6-morpholin-4-yl pyridin-2-yl methanamine
PubChem CID 18525740
SMILES CNCC1=NC(=CC=C1)N1CCOCC1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name methyl({[6-(morpholin-4-yl)pyridin-2-yl]methyl})amine
Formula Weight 207.28
Percent Purity 90%
Chemical Name or Material N-Methyl-N-[(6-morpholin-4-ylpyridin-2-yl)methyl]amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.