missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-{[5-(pyridin-3-yloxy)-2-furyl] methyl}amine, 97%, Thermo Scientific™

Product Code. 10610335
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10610335 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10610335 Supplier Thermo Scientific Maybridge Supplier No. CC42746CB
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 859850-77-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H12N2O2
MDL Number MFCD07772853
Quantity 250 mg
Synonym n-methyl-n-5-pyridin-3-yloxy-2-furyl methyl amine, methyl 5-pyridin-3-yloxy furan-2-yl methyl amine, n-methyl-1-5-pyridin-3-yloxy furan-2-yl methanamine, methyl 5-3-pyridyloxy 2-furyl methyl amine, 2-furanmethanamine,n-methyl-5-3-pyridinyloxy, n-methyl-1-5-pyridin-3-yloxyfuran-2-yl methanamine, n-methyl-1-5-pyridin-3-yl oxy furan-2-yl methanamine
InChI Key MUKCVLFJCGZPKF-UHFFFAOYSA-N
SMILES CNCC1=CC=C(O1)OC2=CN=CC=C2
IUPAC Name N-methyl-1-(5-pyridin-3-yloxyfuran-2-yl)methanamine
Molecular Weight (g/mol) 204.229
PubChem CID 7164635
Formula Weight 204.23
Percent Purity 97%
Chemical Name or Material N-methyl-N-{[5-(pyridin-3-yloxy)-2-furyl]methyl}amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.