missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-((4-methyl-2-phenyl-1,3-thiazol-5-yl)methyl)amine, 97%, Thermo Scientific™

Product Code. p-3111066
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10661465 250 mg 250mg
10671465 1 g 1g
10442824 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10661465 Supplier Thermo Scientific Maybridge Supplier No. CC29246CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 850375-02-9
Molecular Formula C12H14N2S
Molecular Weight (g/mol) 218.318
MDL Number MFCD07368511
InChI Key GFAGRBRYZWAUSV-UHFFFAOYSA-N
Synonym n-methyl-n-4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-thiazol-5-yl methanamine, methyl 4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, 5-thiazolemethanamine,n,4-dimethyl-2-phenyl, n-methyl-1-4-methyl-2-phenylthiazol-5-yl methanamine, methyl 4-methyl-2-phenyl 1,3-thiazol-5-yl methyl amine, mmj
PubChem CID 4961253
IUPAC Name N-methyl-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)methanamine
SMILES CC1=C(SC(=N1)C2=CC=CC=C2)CNC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 218.32
Percent Purity 97%
Chemical Name or Material N-methyl-N-[(4-methyl-2-phenyl-1, 3-thiazol-5-yl)methyl]amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.