missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[4-(2-morpholin-4-ylethoxy)benzyl]amine, 95%, Thermo Scientific™

Product Code. 10739403
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-77-9
Molecular Formula C14H22N2O2
Molecular Weight (g/mol) 250.342
MDL Number MFCD07368554
InChI Key CFDWJPMGCABITD-UHFFFAOYSA-N
Synonym n-methyl-n-4-2-morpholin-4-ylethoxy benzyl amine, methyl 4-2-morpholin-4-yl ethoxy phenyl methyl amine, benzenemethanamine,n-methyl-4-2-4-morpholinyl ethoxy, n-methyl-1-4-2-morpholin-4-ylethoxy phenyl methanamine, n-methyl-n-4-2-morpholin-4-ylethoxy benzylamine, n-methyl-1-4-2-morpholinoethoxy phenyl methanamine, methyl 4-2-morpholin-4-ylethoxy phenyl methyl amine
PubChem CID 7060587
IUPAC Name N-methyl-1-[4-(2-morpholin-4-ylethoxy)phenyl]methanamine
SMILES CNCC1=CC=C(C=C1)OCCN2CCOCC2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 250.34
Percent Purity 95%
Chemical Name or Material N-Methyl-N-[4-(2-morpholin-4-ylethoxy)benzyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'