missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(3-thien-2-ylbenzyl)amine, 97%, Thermo Scientific™

Product Code. 10594994
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10594994 1 g Amber Glass Bottle 1g
10185152 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10594994 Supplier Thermo Scientific Maybridge Supplier No. CC44546DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 859833-20-8
Molecular Formula C12H13NS
Molecular Weight (g/mol) 203.30
MDL Number MFCD08435869
InChI Key BIULJOFRQMLCQK-UHFFFAOYSA-N
Synonym n-methyl-n-3-thien-2-ylbenzyl amine, methyl 3-thiophen-2-yl phenyl methyl amine, methanamine,n-3-2-thienyl phenyl methylene, n-methyl-1-3-thiophen-2-yl phenyl methanamine, n-methyl-n-3-then-2-ylbenzyl amne, methyl 3-2-thienyl phenyl methyl amine, n-methyl-1-3-thien-2-ylphenyl methylamine
PubChem CID 18525772
SMILES CNCC1=CC=CC(=C1)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name methyl({[3-(thiophen-2-yl)phenyl]methyl})amine
Formula Weight 203.31
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(3-thien-2-ylbenzyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.