missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(3-pyridylmethyl)amine, 97%, Thermo Scientific™

Product Code. 10069541
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 20173-04-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00023610
InChI Key MCSAQVGDZLPTBS-UHFFFAOYSA-N
Synonym n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine
PubChem CID 88393
IUPAC Name N-methyl-1-pyridin-3-ylmethanamine
SMILES CNCC1=CN=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material N-methyl-N-(3-pyridylmethyl)amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'