missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-n-(3-piperidin-1-ylbenzyl)amine, 90%, Thermo Scientific™

Product Code. 10411873
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10411873 250 mg Amber Glass Bottle 250mg
10412063 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10411873 Supplier Thermo Scientific Maybridge Supplier No. CC34046CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 859850-65-0
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.317
MDL Number MFCD07772817
InChI Key WHELHMLNRPDTJU-UHFFFAOYSA-N
Synonym n-methyl-n-3-piperidin-1-ylbenzyl amine, methyl 3-piperidin-1-yl phenyl methyl amine, methyl 3-piperidylphenyl methyl amine, benzenemethanamine,n-methyl-3-1-piperidinyl, n-methyl-1-3-piperidin-1-ylphenyl methanamine, n-methyl-1-3-piperidin-1-yl phenyl methanamine
PubChem CID 7162073
IUPAC Name N-methyl-1-(3-piperidin-1-ylphenyl)methanamine
SMILES CNCC1=CC(=CC=C1)N2CCCCC2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.32
Percent Purity 95%
Chemical Name or Material N-methyl-N-(3-piperidin-1-ylbenzyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.