missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide, 97%, Thermo Scientific™

Product Code. 10531123
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10531123 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10531123 Supplier Thermo Scientific Maybridge Supplier No. SB02011EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 108825-65-6
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C15H15N5O
MDL Number MFCD01567008
Quantity 10 g
Synonym n-methyl-n-3-3-methyl 1,2,4 triazolo 4,3-b pyridazin-6-yl phenyl acetamide, acetamide,n-methyl-n-3-3-methyl-1,2,4-triazolo 4,3-b pyridazin-6-yl phenyl, n-methyl-n-3-3-methyl-1,2,4 triazolo 4,3-b pyridazin-6-yl phenyl acetamide, maybridge3_006077, unii-ws1r0n3h0n, acmc-1c5o3, ws1r0n3h0n, acetamide, n-methyl-n-3-3-methyl-1,2,4-triazolo 4,3-b pyridazin-6-yl phenyl
InChI Key WPAQLESUVYGUJZ-UHFFFAOYSA-N
SMILES CC1=NN=C2N1N=C(C=C2)C3=CC(=CC=C3)N(C)C(=O)C
IUPAC Name N-methyl-N-[3-(3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide
Molecular Weight (g/mol) 281.319
PubChem CID 2737312
Formula Weight 281.32
Percent Purity 97%
Chemical Name or Material N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.