missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-L-prolinol, 96%, Thermo Scientific™

Product Code. 11916741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11916741 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11916741 Supplier Thermo Scientific Acros Supplier No. 442530050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 34381-71-0
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 67°C to 69°C (12.0 Torr)
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Molecular Formula C6H13NO
Refractive Index 1.4692
MDL Number MFCD00011727
Quantity 5 g
Specific Gravity 0.97
Specific Rotation -46° to -54° (20°C, 589nm) (c=5 in methanol)
Synonym n-methyl-l-prolinol, s---1-methyl-2-pyrrolidinemethanol, n-methylprolinol, 2s-1-methylpyrrolidin-2-yl methanol, s-1-methylpyrrolidin-2-yl methanol, 2-pyrrolidinemethanol, 1-methyl-, 2s, 2s-1-methyl-2-pyrrolidinyl methanol, s---2-hydroxymethyl-1-methylpyrrolidine, 1-methyl-2-pyrrolidinyl methanol #, n-methyl prolinol
Solubility Information Solubility in water: in water:(20°C) insoluble.,in ethanol:(20°C) soluble
InChI Key VCOJPHPOVDIRJK-UHFFFAOYNA-N
SMILES CN1CCCC1CO
IUPAC Name [(2S)-1-methylpyrrolidin-2-yl]methanol
Molecular Weight (g/mol) 115.18
PubChem CID 643492
Formula Weight 115.18
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material N-Methyl-L-prolinol
Show More Show Less
compliance-icons
Product Identifier
  • N-Methyl-L-prolinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.