missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-DL-alanine, 98%

Product Code. 15442509
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442509 100 g 100g
15432509 25 g 25g
15452509 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442509 Supplier Thermo Scientific Alfa Aesar Supplier No. H65287.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 600-21-5 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-21-5
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00063136
InChI Key GDFAOVXKHJXLEI-UHFFFAOYSA-N
Synonym n-methyl-dl-alanine, 2-methylamino propanoic acid, h-n-me-dl-ala-oh, n-alpha-methyl-dl-alanine, n-me-dl-ala-oh, alanine, n-methyl-, l, dl-n-methylalanine, me-dl-ala-oh, n-methyl cdl-alanine, n-, a-methyl-dl-alanine
PubChem CID 4377
IUPAC Name 2-(methylamino)propanoic acid
SMILES CC(C(=O)O)NC

Specifications

Color White
Quantity 100 g
Formula Weight 103.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-DL-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.