missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(diphenylmethyl)amine, 98%

Product Code. 11489490
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489490 5 g 5g
11479490 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489490 Supplier Thermo Scientific Alfa Aesar Supplier No. H50327.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14683-47-7 | C14H15N | 197.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14683-47-7
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00467853
InChI Key SHDMMLFAFLZUEV-UHFFFAOYSA-N
Synonym n-diphenylmethyl methylamine, methylbenzhydrylamine, n-benzhydryl-n-methylamine, n-methylbenzhydrylamine, diphenylmethyl methyl amine, diphenyl-methyl methyl amine, diphenylmethyl methylamine, n-methyldiphenylmethanamine, acmc-20akaj, n-methyl benzhydrylamine
PubChem CID 411467
IUPAC Name N-methyl-1,1-diphenylmethanamine
SMILES CNC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 38°C to 42°C
Quantity 5 g
Beilstein 2096409
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material N-Methyl(diphenylmethyl)amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.